Anti-FRMD5 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-8237R
BOSSBS-8237REA 719 EUR
BOSSBS-8237R
Anti-FRMD5 Rabbit Polyclonal Antibody
Antikörper
FERM domains are roughly 150 amino acids in length and are found in a number of cytoskeletal-associated proteins such as Ezrin, Radixin, Moesin and 4.1 (erythrocyte membrane protein band 4.1), where they provide a link between cytoskeletal signals and membrane dynamics. FRMD5 (FERM domain-containing protein 5) is a 570 amino acid single-pass membrane protein that contains one FERM domain and exists as two alternatively spliced isoforms. The gene encoding FRMD5 maps to human chromosome 15, which houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FRMD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR