Hand cleanser dispensers, Plus
Lieferant: B. Braun
1418894 1418895 1418892 1418893
BRAU1418894EA 180 EUR
BRAU1418894 BRAU1418895 BRAU1418892 BRAU1418893
Hand cleanser dispensers, Plus
Seifen- und Flüssigseifenspender
Wall mounted hand cleanser dispensers for contact free hygiene applications.
  • Control lever of two different lengths
  • Easy and fast pump cleaning by front removal
  • Flexible seal cap to avoid evaporation and two interchangeable clips - disinfectant/cleanser
Suitable for use with 'standard' 500 and 1000 ml containers.