Alkaline cleaners for machine washing of surgical instruments, Helimatic® Cleaner alcaline
Lieferant: B. Braun
18731
BRAU18731EA 135 EUR
BRAU18731
Alkaline cleaners for machine washing of surgical instruments, Helimatic® Cleaner alcaline
Reinigungsmittel Waschmittel und Oberflächenreiniger
  • Effective cleaner of organic deposits
  • Typical dosage: 0,3 to 0,5%
  • Alkaline> pH10