One-way stopcock, Discofix®
Lieferant: B. Braun
4099117
BRAU4099117EA 196 EUR
BRAU4099117
One-way stopcock, Discofix®
Hähne
Suitable for all pressure infusion systems.
  • Continuous flow channels for accurate pressure monitoring
  • Swivel lock can be moved axially or radially for fast and secure connection
  • PVC- and latex-free