Infusion filter, Sterifix® Neonat
Lieferant: B. Braun
4099257
BRAU4099257EA 19 EUR
BRAU4099257
Infusion filter, Sterifix® Neonat
Infusionen
Infusion filter with bacteria and particle retention.
  • Reliable air venting regardless of position
  • High throughput and Supor® membrane
  • Pressure resistant to one bar