Infusion filter, Intrapur® lipid
Lieferant: B. Braun
4099702
BRAU4099702EA 1900 EUR
BRAU4099702
Infusion filter, Intrapur® lipid
Infusionen
Infusion filter for lipid emulsions and all-in-one parenteral nutrition.
  • Retention of fungi, spores and particles
  • Reliable air venting regardless of position
  • High throughput via Luer lock fitting