Dispensing pins, MICRO PIN®
Lieferant: B. Braun
415019
BRAU415019EA 239 EUR
BRAU415019
Dispensing pins, MICRO PIN®
Infusionen
Single use dispensing pin is used for withdrawal or injection of medication from rubber-stopper vials and bag injection ports.
  • Compatible with Luer lock connection
  • Simple design with large internal lumen
  • Eliminates the risk of needle stick injuries
  • High fluid-flow rate and comfortable easy handling
Made of plastic and free from PVC, DEHP and natural rubber latex.