Thermal transfer printer ribbon, 4300 series, black
Lieferant: Brady
010615
BRDY010615EA 52 EUR
BRDY010615
Thermal transfer printer ribbon, 4300 series, black
Farbbänder
Ribbons for desktop printers and printer applicators.
  • Ribbons use heat and pressure to transfer images
  • Designed for use with desktop or larger printers
Strong environmental resistance against smearing and chemicals. It offers reliable, sharp-edge print quality to maximise bar code readability.