Labels for thermal transfer printers, type B-438
Lieferant: Brady
030182
BRDY030182EA 229.37 EUR
BRDY030182
Labels for thermal transfer printers, type B-438
Etiketten
  • Fuels or oils resistant
  • High and low temperatures resistant
  • Solvents or chemicals resistant
  • Fraud resistant
Standards: UL recognised.