Thermal transfer printer ribbon, R4307 series
Lieferant: Brady
030253
BRDY030253EA 243.47 EUR
BRDY030253
Thermal transfer printer ribbon, R4307 series
Farbbänder
Strong environmental resistance against smearing and chemicals. Also, reliable, sharp-edge print quality to maximise bar code readability.
  • Core - diameter (mm): 25.40 mm
  • Ribbon formulation: wax/resin
  • Ink side of ribbon: ink side out
Verpackung: One ribbon(s)/box.