Lockout hasp, non conductive
Lieferant: Brady
045582
BRDY045582EA 12.55 EUR
BRDY045582
Lockout hasp, non conductive
Sicherheitsverriegelungen
These non conductive lockout hasps have exceptional electrical insulating characteristics and are made of strong, rigid red plastic resin.
  • High flexibility
  • High resistance to cracking and most chemicals
Max. shackle diameter: 9,00 mm
Min. shackle clearance: 20,00 mm
Shackle diameter: 6,10 mm