Gas cylinder lockout
Lieferant: Brady
045629
BRDY045629EA 64.37 EUR
BRDY045629
Gas cylinder lockout
Sicherheitsverriegelungen
This lockout made of red PP prevents access to the main cylinder valve. It is manufactured to accommodate neck rings up to 88 mm in diameter.
  • No need to match the lockout device to varying diameters and threads of cylinder caps
  • Gas cylinder lockout is adjustable and installs in seconds
Max. shackle diameter: 7 mm
Min. shackle clearance: 20 mm
Hole diameter: 36 mm