Labels for 76 mm Core Thermal Transfer Printers, Type B-427
Lieferant: Brady
062369 030349 030536 030538 062333 062361 062367 030112 030539 030249 030233 062364 030113 151022
BRDY062369EA 343.42 EUR
BRDY062369 BRDY030349 BRDY030536 BRDY030538 BRDY062333 BRDY062361 BRDY062367 BRDY030112 BRDY030539 BRDY030249 BRDY030233 BRDY062364 BRDY030113 BRDY151022
Labels for 76 mm Core Thermal Transfer Printers, Type B-427
Etiketten
Printable, self-laminating vinyl labels (B-427) for thermal transfer printers. For smooth surfaces.
  • Self-extinguishing
  • Excellent water and oil resistance
  • Excellent abrasion and smudge resistance
Ideal for barcode labelling, component and equipment labelling, general identification, inventory and inspection labelling.
Ribbon series:
Standards: This material is UL recognised with its respective ribbon.