Plug lockout, Stopower™
Lieferant: Brady
065673
BRDY065673EA 23.5 EUR
BRDY065673
Plug lockout, Stopower™
Sicherheitsverriegelungen
This plug hugger is ideal for office equipment and other frequently locked out cords.
  • For use on 2 or 3 pronged polarised plugs
  • Cord cannot be plugged in with Stopower in place
  • Device cannot be removed without a key