Snap-on breaker lockout, EZ Panel Loc™
Lieferant: Brady
051252 065815
BRDY051252EA 30.46 EUR
BRDY051252 BRDY065815
Snap-on breaker lockout, EZ Panel Loc™
Sicherheitsverriegelungen
The snap-on breaker is for use on Square D and other breakers that have a hole in the switch tongue.
  • Just align pins with hole and snap closed
  • Pins prevent switch from being thrown
  • Made from high-impact isoplast polymers