Signs, floor signs, ToughStripe™ , dots
Lieferant: Brady
104400
BRDY104400EA 252.46 EUR
BRDY104400
Signs, floor signs, ToughStripe™ , dots
Warn- und Verbotszeichen
ToughStripe™ products are used for marking aisles, passageways and storage locations, and pointing out important safety equipment and egress paths.
  • Floor marking products are made with B-514 polyester
  • Recommended for indoor use and dry environment
  • Resistant to harsh chemicals