Polyester Tags
Lieferant: Brady
105370 105371
BRDY105370EA 60.34 EUR
BRDY105370 BRDY105371
Polyester Tags
Etiketten
These two part tags provide enhanced operational control and record keeping. Matching identification numbers are printed on each part.
  • Write-on surface accepts marker, pen or pencil
  • Weatherproof for either indoor or outdoor use
  • Employs ³/₈" rolled brass eyelet to fit over padlock shackles
Polyester 10 mm thickness enhances durability to easily remove chemicals, grease and grime.
Lieferumfang: Nylon cable ties included for easy and secure attachment.