Labels for BBP85 printers, type B-7569
Lieferant: Brady
120974 120989
BRDY120974EA 163.85 EUR
BRDY120974 BRDY120989
Labels for BBP85 printers, type B-7569
Etiketten
B-7569 is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
  • Gloss finish
  • Thermal transfer print technology
Ideal for facility identification, floor marking, hazard communication identification, safety identification, warehouse marking.
Standards: RoHS compliant with Directive 2011/65/EU and 2015/863, Part does not contain an SVHC, S5609:1986 Section 2 and section 3 approval/compliance.