Cable ties, stainless steel
Lieferant: Brady
134057
BRDY134057EA 41.75 EUR
BRDY134057
Cable ties, stainless steel
Kabelbinder
Brady B-7316 stainless steel cable ties are made from 316 grade stainless steel and are designed to withstand harsh environments, and offer excellent outdoor durability.
  • Thickness of 0,5 mm