Preprinted Tags "Energy Source, Hydraulic"
Lieferant: Brady
138410 138411 138412 138744 138745 138746 138747 138748 138749 138750
BRDY138410EA 57.91 EUR
BRDY138410 BRDY138411 BRDY138412 BRDY138744 BRDY138745 BRDY138746 BRDY138747 BRDY138748 BRDY138749 BRDY138750
Preprinted Tags "Energy Source, Hydraulic"
Etiketten
These hydraulic energy source tags are constructed of PP with round corners with one hole.
  • Permanent acrylic adhesive
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Colour: Olive green on white
Standards: ATEX
Verpackung: 25 tags per pack.