Preprinted Tags "Energy Source, Thermal"
Lieferant: Brady
138434 138437 138438 138780 138781 138782 138783 138784 138785 138786
BRDY138434EA 57.91 EUR
BRDY138434 BRDY138437 BRDY138438 BRDY138780 BRDY138781 BRDY138782 BRDY138783 BRDY138784 BRDY138785 BRDY138786
Preprinted Tags "Energy Source, Thermal"
Etiketten
These thermal energy source tags are constructed of PP with round corners with one hole.
  • Permanent acrylic adhesive
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Colour: Green on white
Standards: ATEX
Verpackung: 25 tags per pack.