Labels for BBP33 printers, type B-7642, PermaSleeve® HX
Lieferant: Brady
142905 142907 142908 142910 142897 142898 142899 142900 142901 142904 142903 142902 142914 142915
BRDY142905EA 237.17 EUR
BRDY142905 BRDY142907 BRDY142908 BRDY142910 BRDY142897 BRDY142898 BRDY142899 BRDY142900 BRDY142901 BRDY142904 BRDY142903 BRDY142902 BRDY142914 BRDY142915
Labels for BBP33 printers, type B-7642, PermaSleeve® HX
Etiketten
PermaSleeve® HX Polyolefin wire marker sleeves are military/industry grade.
  • Flame-retardant
  • Temperature resistance from –40 to 120 °C
Standards: Meets SAE-AMS-DTL-23053/5 (class 1 and 3), SAE-AS-81531- MIL-STD-202 and tested ASTM E162, ASTM E662.