Labels for BBP33 printers, type B-343, PermaSleeve® PS
Lieferant: Brady
143036 143033 143034 143035 143037 143038 143039 143040 143026 142988 142990 142992 142993 142989 142944 142945 142946 142934 142921 143041 142937 142923 142926 142927 142928 142929 142930 142931 142932 142933 143028 143019 143020 143021 143022 143023 143024 143025 143027 143029 143030 143031 143032 133809 133879 133978 133979 133784 133785
BRDY143036EA 565.55 EUR
BRDY143036 BRDY143033 BRDY143034 BRDY143035 BRDY143037 BRDY143038 BRDY143039 BRDY143040 BRDY143026 BRDY142988 BRDY142990 BRDY142992 BRDY142993 BRDY142989 BRDY142944 BRDY142945 BRDY142946 BRDY142934 BRDY142921 BRDY143041 BRDY142937 BRDY142923 BRDY142926 BRDY142927 BRDY142928 BRDY142929 BRDY142930 BRDY142931 BRDY142932 BRDY142933 BRDY143028 BRDY143019 BRDY143020 BRDY143021 BRDY143022 BRDY143023 BRDY143024 BRDY143025 BRDY143027 BRDY143029 BRDY143030 BRDY143031 BRDY143032 BRDY133809 BRDY133879 BRDY133978 BRDY133979 BRDY133784 BRDY133785
Labels for BBP33 printers, type B-343, PermaSleeve® PS
Etiketten
PermaSleeve® PS heat-shrink polyolefin (B-342) wire marking sleeves combine speed, ease of use, and permanence to create long-lasting, high quality identification.
  • Fade-resistant and smudge-proof for superior aesthetic appearance
  • Cold resistant and flame-retardant with an operating temperature of –55 to +135 °C
3:1 heat shrinkable sleeve fit snugly around wires for maximum insulation protection and permanent identification. 25 mm sleeve width comes with '2-up pre-cut' configuration.
Standards: Meets SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202; RoHS Compliant and UL recognised.