Ribbons for label printer BBP™33
Lieferant: Brady
143118
BRDY143118EA 80.14 EUR
BRDY143118
Ribbons for label printer BBP™33
Farbbänder
'Drop & Click' print ribbons for quick and easy supply changes.
  • Tough, smear-proof ribbons for long-lasting printed text
  • Print can withstand years in industrial areas and outdoor conditions
  • No calibrating, no threading, no waste, no hassle
Width: 104,39 mm