Workstation lockout writer software app
Lieferant: Brady
145445
BRDY145445EA 832.99 EUR
BRDY145445
Workstation lockout writer software app
Software Labor-Software
Workstation lockout writer app allows you to create compliant lockout/tagout labels, tags and procedures in no time.
  • It has printing templates, simple translations, easy-to-use image editing for callouts, and more
  • Features drop down menus for navigation between hazards, energy sources and devices to help you create custom standards for your procedures
  • Helps with OSHA standards compliance for the lockout of hazardous energy sources
  • Includes import capabilities from Lockout-Pro® software
  • Minimum 1 GB RAM
  • 200 MB free desk space required
Operating system: Windows® 10;Windows® 7 (32- or 64-bit);Windows® 8 (32- or 64-bit);Windows® 8.1 (32- or 64-bit).