Brady® SPC® Absorbents HAZWIK Quick Response Kit, Chemical
Lieferant: Brady
152186
BRDY152186EA 220.44 EUR
BRDY152186
Brady® SPC® Absorbents HAZWIK Quick Response Kit, Chemical
Absorptionsmaterial
Get fast and effective spill cleanup with a HAZWIK® quick response kit. It contains everything you need for emergency spills in a convenient, grab-and-go-style bag.
  • Compact, single-use kit that is ideal for use on small spills in the laboratory
  • Quickly cleans up hazardous chemical spills
  • Visual instructions on the package
  • Contains PPE, tools, absorbent pads, acid and base neutralisers, wipes and disposable materials
Lieferumfang: Includes 0,13 L acid neutraliser (1), 0,13 L base neutraliser (1), cable tie (1), disposal bag (1), pair of nitrile gloves (1), scoop and scraper (1), surface sanitising wipe (1), 190,5×254 mm pads (6).
Verpackung: Packed as five individual kits per case.