Brady® SPC® Absorbents QUICKWIK Universal Granular Absorbent
Lieferant: Brady
152275
BRDY152275EA 1939.98 EUR
BRDY152275
Brady® SPC® Absorbents QUICKWIK Universal Granular Absorbent
Absorptionsmaterial
Take care of industrial spills, leaks and drips with QUICKWIK™ universal granular absorbent – more than 4× the absorption of clay granular and produces no carcinogens or dust.
  • Premium absorbent made of 100% recycled material
  • Effectively absorbs hydrocarbons, water-based fluids and most aggressive chemicals
  • Certified safe for landfill disposal
  • Lightweight and easy to deploy
Verpackung: Supplied in 50 litres bags, 48 bags per pallet.