BradyJet J4000-EU Label Printer, Colour
Lieferant: Brady
152711
BRDY152711EA 3286.06 EUR
BRDY152711
BradyJet J4000-EU Label Printer, Colour
Drucker Label Printers
High-impact, precision full colour that lasts. Create the compliant, long-lasting labels, signs and tags you need with the BradyJet J4000 inkjet printer.
  • Full colour
  • Long-lasting
  • 5000 labels per day
This industry inkjet workhorse creates up to 5000 impactful full-colour labels per day up to 203,20 mm wide on continuous or die-cut material. Ideal for a wide variety of applications in safety, including lean/5S, lockout tagout, instructional or safety chemical labelling for handling, storage or transport, as well as lab settings. Contains the colour options you need to create ANSI, GHS, ISO, BS5609 and arc flash compliant labels.
Streamline storage by only needing to stock white label material and print only what you need, when you need it, in photo-quality full colour every time. Easily print PDFs or from third-party software packages with the optimised windows driver. Add Brady workstation safety and facility ID doftware suite (J4000-BWSSFID) or lab suite (J4000-BWSLAB) to your solution and make label creation even easier.
Standards: CE; C-UL US; FCC Class A; RoHS directive 2011/65/EU; UL Listed for US.
Lieferumfang: Includes (1) BradyJet J4000 printer; (1) power cord; (1) CMY ink cartridge with built in printhead; (1) pack of 2 ink cartridge storage caps; (1) media starter roll 101,60 mm wide B-2595 vinyl; (1) USB cord; (1) USB flash drive with product information; (1) Brady workstation basic suite.