Self-laminating Polyester Cryogenic Laboratory Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Lieferant: Brady
170770
BRDY170770EA 56.47 EUR
BRDY170770
Self-laminating Polyester Cryogenic Laboratory Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketten
Securely label tubes and vials for cryogenic storage with this self-laminating, ultra-thin polyester label material (B-461). Print almost anywhere with a compact Brady portable label printer.
  • Ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws, and slides
  • Able to withstand extreme temperatures from –196 to +130 °C, immersion in liquid nitrogen, and autoclave use
  • Resistant to ethanol, toluene, isopropanol, xylene, DMSO, 50% acetic acid, 10% hydrochloric acid, 10% Clorox solution
  • Made with a clear overlaminate that protects the print and maintains content visibility
  • Matte finish
Matte finish, chemical-resistant, low temperature-resistant, smear-resistant B-461 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
Standards: RoHS compliant to 2011/65/EU (Pass 2015/863).
Verpackung: Labels are packaged 120 per box.