Solvent and Chemical Resistant Polyester Labels with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Lieferant: Brady
170774 170887
BRDY170774EA 48.42 EUR
BRDY170774 BRDY170887
Solvent and Chemical Resistant Polyester Labels with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketten
High-performance polyester (B-488) withstands numerous solvents and variable temperatures when applied to various surfaces, including stainless steel and polypropylene.
  • Specialized – Use B-488 high-performance polyester label material for lab applications with numerous solvents and variable temps
  • Durable – Labels resist chemicals, abrasions, corrosion and humidity and have a high/low temperature performance of –40 to +160 °C (–40 to +320 °F)
  • Adhesive – Made with a permanent acrylic adhesive for tight bonding onto flat, smooth surfaces
Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material.
Ideal for sset tracking, barcode labeling, component and equipment labeling, data and telecommunications labeling, general identification, inventory and inspection labeling, laboratory labeling.
Matte finish, abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant B-488 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
Standards: RoHS compliant to 2011/65/EU and 2015/863.