Labels for BMP™ 51/53 printer, type B-595
Lieferant: Brady
170949
BRDY170949EA 24.79 EUR
BRDY170949
Labels for BMP™ 51/53 printer, type B-595
Etiketten
These are made from durable, low-shrink vinyl with a very aggressive adhesive. The labels adhere to irregular, curved, rough and highly textured surfaces (painted cinder blocks, uneven wood, textured plastics, paper jacketed pipes, powder coated surfaces). Also adhere to difficult low energy surface items (PVC piping, blow-moulded equipment cases, ABS plastics, recycled plastics).