Printer Ribbons, M710 R10000 Series
Lieferant: Brady
173199 173196 173195 173194 173197 173198
BRDY173199EA 159.88 EUR
BRDY173199 BRDY173196 BRDY173195 BRDY173194 BRDY173197 BRDY173198
Printer Ribbons, M710 R10000 Series
Farbbänder
Ideal for marking pipes and valves, facility safety, and compliance ID and general ID.
  • Offers strong solvent and scratch resistance, plus high-performance images
  • Self-contained ribbon cartridge is easy to replace
Featuring a 5 second 'drop and click' supply change, this smear-proof M710 ribbon needs no threading or calibration. And, it’s built super-tough for outdoor and industrial use.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable