High Tensile Polypropylene Inserts/Tags for M610, M611, BMP61, M710 and BMP71
Lieferant: Brady
173456
BRDY173456EA 394.04 EUR
BRDY173456
High Tensile Polypropylene Inserts/Tags for M610, M611, BMP61, M710 and BMP71
Etiketten
Polypropylene tag stock (B-412) is ideal for wire and cable identification and product inventory identification, where legibility and tensile strength are needed.
  • Non-adhesive material is thermal transfer printable
  • Good abrasion and chemical resistance
  • Service temperature of −40 to +38 °C (−40 to +100 °F)
  • Matte finish
Applications include component and equipment labelling, data and telecommunications labelling, electrical labelling, and wire and cable labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6210 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R6200 for the M710/BMP71 label printers.
Verpackung: 5000 labels per box.