Polyethylene Tags for M710, BMP71, M610, BMP61, and M611 Printers
Lieferant: Brady
173984
BRDY173984EA 35.85 EUR
BRDY173984
Polyethylene Tags for M710, BMP71, M610, BMP61, and M611 Printers
Etiketten
Multi-purpose tags can be used for a variety of applications, including identification of multi-conductor cables, inventory, equipment, lockout, safety warning repair, and work-in-progress.
  • Cross-laminated polyethylene material
  • Extremely tear-resistant and superior cold-weather tag
  • Rectangular label with oval hole
  • Die-cut
Tags for the M610, BMP61, and M611 thermal transfer portable printers. Please Note: This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use. These parts can also be printed on the BMP™71 label printer and M710 printer provided the roll adapter (M71-ADAPT) is used.
Standards: ASTM D 1000, ASTM D 1938, ASTM G 115, ASTM G 155. RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.