Polypropylene Flag Labels for M710, M610 and M611
Lieferant: Brady
174383 174381
BRDY174383EA 62.63 EUR
BRDY174383 BRDY174381
Polypropylene Flag Labels for M710, M610 and M611
Etiketten
Flags for the M610, M611, and M710 Portable Printers.
  • No visible affect to label when exposed to most common chemicals, oils, or water
  • Service temperature of −70 to 80 °C
Standards: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815).