ToughStripe® Arrows
Lieferant: Brady
175655
BRDY175655EA 68.89 EUR
BRDY175655
ToughStripe® Arrows
Warn- und Verbotszeichen
ToughStripe® Arrows provide visible floor marking for improved safety and organisation in almost any industrial environment.
  • Polyester with polyester overlaminate
  • Permanent rubber adhesive
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Ideal for marking aisles, passageways and storage locations, and identifying equipment and egress paths
  • Can be easily applied by one person working alone
Extremely durable ToughStripe® die-cut floor marking arrows are made of rugged polyester with ultra-aggressive adhesive and hold up against forklift traffic.
Verpackung: 25 labels per pack.