Multicolor and Cut, Sign and Label Printers, S3700
Lieferant: Brady
176427 176428 176429 176430 176431 176432
BRDY176427EA 3449 EUR
BRDY176427 BRDY176428 BRDY176429 BRDY176430 BRDY176431 BRDY176432
Multicolor and Cut, Sign and Label Printers, S3700
Drucker Label Printers
Color your way to an improved workplace with the S3700 Sign and Label Printer. With multicolor print and shape cutting capabilities, this printer creates impactful identification that fits your application and workspace needs.
  • Color printing: Multicolor, single color
  • Material: Plastic
  • Warranty: one year
  • Multicolor print capabilities make it ideal for creating impactful safety signs, lockout tags, warning labels and more in a single pass
  • Built in XY-cutter allows for the creation of die-cut shapes and text that gives the flexibility to choose where to place
More Product Information