Work smarter. Label larger with the M511. Print bigger, clearer messages. Go edge to edge. Design and print on the fly, or a PC. Plus, share files, import data and connect up to 5 users simultaneously.
- Monocolour
- Ideal for larger applications - print up to 38,10 mm labels
- Less waste - go edge to edge with text, graphics and barcodes
- Longer battery life with an internal Li-ION battery - 8+ hours of work; 1000 labels on a single charge
- More durable - resists 1,8 m drops, shocks, 115 kg crushes and blowing sand and dust (military-grade shock resistance; MIL-STD-810G)
- Prints anywhere - Bluetooth 5.0 (20 m range); connects up to 5 users simultaneously
- Uses label creation software - Express Labels app via Bluetooth on a phone/tablet; Brady Workstation on a PC
- Saves time; saves space - features an auto label cutter and prints upright or on its side
Asset tracking, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, facility identification, general identification, inventory and inspection labelling, laboratory labelling, lean and 5S labelling, safety identification, warehouse marking, wire and cable labelling.
Standards: RoHS 2011/65/EU, 2015/863; CE; UKCA; cULus (US/Canada); FCC.
Lieferumfang: Kit includes Brady Workstation Product and Wire ID software. This printer ships in a hard case with a M511 printer, M5C-1500-595-WT-BK, M4-91-427, M5-131-499 label cartridges, an AC Adaper, a magnet, a utility hook, a Power Brick for M211 and M511 printers, a Quick Start Guide and Brady Workstation Product and Wire ID Software Suite.