Work the entire day using this AC adapter/battery charger. It powers the M410, M510, M511, M610 and M611 label printers and charges the related battery packs.
- Use this adapter to power and charge your M410, M510, M511, M610 and M611 label printers
- Includes an AC adapter/charger
- Power supply voltage 110 to 240 V
Work the entire day using this AC Adapter/Battery Charger. It powers the M410, M510, M511, M610 and M611 label printers and charges the related battery packs. Use this adapter to power and charge your M410, M510, M511, M610 and M611 label printers.. Includes an AC Adapter/Charger. Power supply voltage: 110 - 240 V.
Bestellinformation: Order reference M41-51-61-AC.
Lieferumfang: AC adapter and power cord.