M410 Rechargeable Battery
Lieferant: Brady
176993
BRDY176993EA 83.41 EUR
BRDY176993
M410 Rechargeable Battery
Batterien
Get longer battery life and save money on AA batteries with this rechargeable NiMH battery for M410, BMP41 and BMP61 Label Printers. Printer can be run while battery is charging.
  • Long battery life - prints up to 1000 labels per battery charge
  • Use AC adapter/charger unit (not included) to charge battery
  • Enables cordless printer operation so you can print labels where you need them, when you need them
Get longer battery life and save money on AA batteries with this rechargeable NiMH battery for M410, BMP41 and BMP61 Label Printers. Printer can be run while battery is charging. Long battery life - prints up to 1000 labels per battery charge. Use AC adapter/charger unit (not included) to charge battery. Enables cordless printer operation so you can print labels where you need them, when you need them.
Bestellinformation: Order reference M410-BATT.
Lieferumfang: Rechargeable NiMH battery.