Brady® Harsh Environment Multi-Purpose Polyester Labels for M410, M510 and M511 Printers
Lieferant: Brady
177006
BRDY177006EA 54.51 EUR
BRDY177006
Brady® Harsh Environment Multi-Purpose Polyester Labels for M410, M510 and M511 Printers
Etiketten
Harsh environment applications require tough materials and Brady's B-423 polyester stands up to the challenge.
  • High-tack permanent adhesive for a variety of surface types in all-weather applications
  • Resists exposure to UV light, extreme temperatures, abrasions, fuel and oil, degreasers, cleaners, and detergents
  • Compatible with Brady printers BMP41, BMP51, BMP53, M410, M510, and M511
  • For use with ribbon series R6200
Brady's B-423 Harsh Environment Multi-Purpose Polyester Labels are designed to handle the rigors of renewable energy applications, as this material is ideal for a wide variety of difficult surfaces in an array of environmental conditions.
Standards: UL Recognised to UL969, CSA to C22.2 No. 0.15-95, RoHS compliant to 2011/65/EU (Pass 2015/863), DIN VDE 0472 Part 815.