Brady® Repositionable Multi-Purpose Vinyl Labels for M410, M510, and M511 Printers
Lieferant: Brady
177008 177009
BRDY177008EA 59.65 EUR
BRDY177008 BRDY177009
Brady® Repositionable Multi-Purpose Vinyl Labels for M410, M510, and M511 Printers
Etiketten
Repositionable vinyl film (B-581) is a durable, flexible, high-gloss material ideal for indoor/outdoor marking where frequent repositioning is required.
  • Vinyl film can be thermal transfer printed
  • Average outdoor durability is 5 years
  • Service temperature of −40 to +82 °C
  • Compatible with Brady printers BMP41, BMP51, BMP53, M410, M510, and M511
  • For use with ribbon series R6000HF
Special aggressive adhesive stays in place, but removes cleanly without leaving adhesive residue, making it ideal for applications like 5S and inventory.