Brady® All Weather Permanent Adhesive Vinyl Labels for M410, M510, and M511 Printers
Lieferant: Brady
177010
BRDY177010EA 88.45 EUR
BRDY177010
Brady® All Weather Permanent Adhesive Vinyl Labels for M410, M510, and M511 Printers
Etiketten
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label printer.
  • Ideal for labelling pipes, equipment, shelves, and other industrial surfaces
  • Made with high-tack, permanent acrylic adhesive
  • Compatible with Brady printers BMP41, BMP51, BMP53, M410, M510, and M511
  • For use with ribbon series R6000HF
Brady's low-shrink vinyl label material (B-595) is designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime. Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures. These weather-resistant labels are designed to last up to 10 years in an outdoor environment.