Temperature Indicating Labels, Go or No Go
Lieferant: Brady
195843
BRDY195843EA 20.76 EUR
BRDY195843
Temperature Indicating Labels, Go or No Go
Temperatur-Indikatorstreifen
Reversible temperature indicating labels are part of the VisAlert® Series from Brady®, which is designed to provide immediate visual indication of water or heat exposure.
  • Large temperature range for applications requiring temperature monitoring between 0 and 100 °C
  • Strong permanent acrylic adhesive for any surface
  • Made of sub-surface printed Polyester
Cost-effective solution for temperature checks during maintenance or for trouble spotting during research and development. Extremely low profile is ideal for components, industrial and household applications in areas where measuring temperature is difficult or impractical.
Colour: Green, Red