Spill workfloor
Lieferant: Brady
196262
BRDY196262EA 733.74 EUR
BRDY196262
Spill workfloor
Absorptionsmaterial
Workfloors are a great way to handle and store hazardous fluids and contain potentially hazardous leaks in storage and dispensing areas.
  • Can hold/carry drums or smaller containers
  • Can be easily assembled with other units by using the connector system
  • Compatible with most forklifts and pallet trucks