Label printer tags for BBP33/i3300 Printers, type B-7643
Lieferant: Brady
197481 197482 197483 197484 197485 197486 197487 197488 197489 197490 197491 197492 197493 197494 197495 197496 197497 197498 197499 197500 197501 197503 197504 197505 197506 197507 197508 197509
BRDY197481EA 368.78 EUR
BRDY197481 BRDY197482 BRDY197483 BRDY197484 BRDY197485 BRDY197486 BRDY197487 BRDY197488 BRDY197489 BRDY197490 BRDY197491 BRDY197492 BRDY197493 BRDY197494 BRDY197495 BRDY197496 BRDY197497 BRDY197498 BRDY197499 BRDY197500 BRDY197501 BRDY197503 BRDY197504 BRDY197505 BRDY197506 BRDY197507 BRDY197508 BRDY197509
Label printer tags for BBP33/i3300 Printers, type B-7643
Etiketten
These B-7643 cable tags offers excellent fire safety characteristics combined with minimal smoke emission.
  • No added halogens
  • Made of thermoplastic polyether polyurethane material
Ideal for data and telecommunications labelling, wire and cable labelling.
Standards: RoHS compliance with Directive 2011/65/EU and 2015/863. Halogen free per IEC 61249-2-21 (2003-11), Adherence: SAE-AS81531-1998, Flame retardant: MIL-STD-202G.