Industrial label printers, BradyPrinter i7100
Lieferant: Brady
149047 198613 149046
BRDY149047EA 2142 EUR
BRDY149047 BRDY198613 BRDY149046
Industrial label printers, BradyPrinter i7100
Drucker Label Printers
The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing on lots of different materials within the Aerospace, Defence and Mass Transit, Electronics, Automotive, Logistics, Electrical, Voice-Datacom and LAB markets.
  • 300 or 600 dpi
  • Applications include cable and wire identification, circuit board and component identification, and general and industrial identification
  • Mono colour
  • Print speed upto 300 mm/s
  • Powerful internal processor designed to handle up to 7000 labels per day