Thermal transfer printer labels, type B-8423
Lieferant: Brady
236719 236715 236713 236717 236701 062397 062399 236707 062400 217053 236705 236709
BRDY236719EA 103.98 EUR
BRDY236719 BRDY236715 BRDY236713 BRDY236717 BRDY236701 BRDY062397 BRDY062399 BRDY236707 BRDY062400 BRDY217053 BRDY236705 BRDY236709
Thermal transfer printer labels, type B-8423
Etiketten
Printable satin polyester labels for thermal transfer printers.
  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.