Economy lockout
Lieferant: Brady
236920
BRDY236920EA 9.83 EUR
BRDY236920
Economy lockout
Sicherheitsverriegelungen
This economy lockout is ideal for use on fuse or switch boxes.
  • Made from sturdy steel
  • Six padlocks can be applied
Max. shackle diameter: 10,00 mm
Min. shackle clearance: 20,00 mm