Scissor-lok lockout devices
Lieferant: Brady
236921
BRDY236921EA 78.45 EUR
BRDY236921
Scissor-lok lockout devices
Sicherheitsverriegelungen
These Scissor-lok devices lock out multiple gate valves simultaneously.
  • Secure power cords, freestanding equipment, tanks, cylinders or safety guards
  • Three padlocks can be applied
  • When using own cable, the outside diameter must be 4,76 mm or less
  • Made from moulded glass-filled nylon
  • With vinyl coated cable (0,9 or 3 m long)
Minimum shackle clearance: 19,05 mm
Maximum shackle diameter: 7,00 mm
Max. shackle diameter: 7,00 mm